accessibility menu, dialog, popup

UserName

CAS RN 176650-93-4

IUPAC Name: 1-{[3-(bromomethyl)-5-[(3,5-dimethoxyphenyl)methoxy]phenoxy]methyl}-3,5-dimethoxybenzene
Synonyms: 1-[3-(bromomethyl)-5-[(3,5-dimethoxyphenyl)methoxy]phenoxymethyl]-3,5-dimethoxybenzene
Molecular Weight (g/mol): 503.39
Molecular Formula: C25H27BrO6
InChi Key: RLACLNXCSSBIKS-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(COC2=CC(OCC3=CC(OC)=CC(OC)=C3)=CC(CBr)=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results