missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis(3,5-dimethoxybenzyloxy)benzyl Alcohol 94.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 440.49 |
| Percent Purity | ≥94.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,5-Bis(3,5-dimethoxybenzyloxy)benzyl Alcohol |
Safety and Handling
| TSCA | No |