accessibility menu, dialog, popup

UserName

CH3OOOHOOCH3OCH3OCH3

CAS RN 176650-92-3

IUPAC Name: {3,5-bis[(3,5-dimethoxyphenyl)methoxy]phenyl}methanol
Synonyms: {3,5-bis[(3,5-dimethoxyphenyl)methoxy]phenyl}methanol
Molecular Weight (g/mol): 440.49
Molecular Formula: C25H28O7
InChi Key: TWHQMXZJXJJPKN-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(COC2=CC(OCC3=CC(OC)=CC(OC)=C3)=CC(CO)=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results