missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(3,5-dimethoxybenzyloxy)benzyl Alcohol 94.0+%, TCI America™

Catalog No. B22461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22461G 1 g
B22465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22461G Supplier TCI America Supplier No. B22461G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 176650-92-3
Molecular Formula C25H28O7
Molecular Weight (g/mol) 440.49
MDL Number MFCD03844779
InChI Key TWHQMXZJXJJPKN-UHFFFAOYSA-N
PubChem CID 15778112
IUPAC Name {3,5-bis[(3,5-dimethoxyphenyl)methoxy]phenyl}methanol
SMILES COC1=CC(OC)=CC(COC2=CC(OCC3=CC(OC)=CC(OC)=C3)=CC(CO)=C2)=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 440.49
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(3,5-dimethoxybenzyloxy)benzyl Alcohol