missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis[3,5-bis(benzyloxy)benzyloxy]benzyl Bromide 97.0+%, TCI America™

Catalog No. B21181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21181G 1 g
B21185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21181G Supplier TCI America Supplier No. B21181G

Chemical Identifiers

CAS 129536-41-0
Molecular Formula C49H43BrO6
Molecular Weight (g/mol) 807.781
MDL Number MFCD02093449
InChI Key WEIQDYRDUFADRJ-UHFFFAOYSA-N
Synonym 3,5-Bis(3,5-dibenzyloxybenzyloxy)benzyl Bromide
PubChem CID 11967226
IUPAC Name 1,3-bis[[3,5-bis(phenylmethoxy)phenyl]methoxy]-5-(bromomethyl)benzene
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2)COC3=CC(=CC(=C3)CBr)OCC4=CC(=CC(=C4)OCC5=CC=CC=C5)OCC6=CC=CC=C6)OCC7=CC=CC=C7

Specifications

Melting Point 129°C
Color White
Quantity 1 g
Formula Weight 807.78
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis[3,5-bis(benzyloxy)benzyloxy]benzyl Bromide
TSCA No
Recommended Storage Refrigerator