accessibility menu, dialog, popup

UserName

OOBrOOOO

CAS RN 129536-41-0

IUPAC Name: 1,3-bis(benzyloxy)-5-[(3-{[3,5-bis(benzyloxy)phenyl]methoxy}-5-(bromomethyl)phenoxy)methyl]benzene
Synonyms: 1,3-bis(benzyloxy)-5-(3-{[3,5-bis(benzyloxy)phenyl]methoxy}-5-(bromomethyl)phenoxymethyl)benzene
Molecular Weight (g/mol): 807.78
Molecular Formula: C49H43BrO6
InChi Key: WEIQDYRDUFADRJ-UHFFFAOYSA-N
SMILES: BrCC1=CC(OCC2=CC(OCC3=CC=CC=C3)=CC(OCC3=CC=CC=C3)=C2)=CC(OCC2=CC(OCC3=CC=CC=C3)=CC(OCC3=CC=CC=C3)=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results