missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis[3,5-bis(3,5-dimethoxybenzyloxy)benzyloxy]benzyl Bromide 97.0+%, TCI America™

Catalog No. b22481g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22481G Supplier TCI America Supplier No. B22481G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 152811-37-5
Molecular Formula C57H59BrO14
Molecular Weight (g/mol) 1047.989
InChI Key BMISSUNAKMQTFS-UHFFFAOYSA-N
PubChem CID 15891038
IUPAC Name 1,3-bis[[3,5-bis[(3,5-dimethoxyphenyl)methoxy]phenyl]methoxy]-5-(bromomethyl)benzene
SMILES COC1=CC(=CC(=C1)COC2=CC(=CC(=C2)COC3=CC(=CC(=C3)CBr)OCC4=CC(=CC(=C4)OCC5=CC(=CC(=C5)OC)OC)OCC6=CC(=CC(=C6)OC)OC)OCC7=CC(=CC(=C7)OC)OC)OC

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 1047.99
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis[3,5-bis(3,5-dimethoxybenzyloxy)benzyloxy]benzyl Bromide
TSCA No
Recommended Storage Refrigerator