accessibility menu, dialog, popup

UserName

CH3OOOBrOOOCH3OCH3OOH3COCH3OOCH3OCH3OCH3

CAS RN 152811-37-5

IUPAC Name: 1-[(3-{[3-({3,5-bis[(3,5-dimethoxyphenyl)methoxy]phenyl}methoxy)-5-(bromomethyl)phenoxy]methyl}-5-[(3,5-dimethoxyphenyl)methoxy]phenoxy)methyl]-3,5-dimethoxybenzene
Synonyms: 1-{3-[3-({3,5-bis[(3,5-dimethoxyphenyl)methoxy]phenyl}methoxy)-5-(bromomethyl)phenoxymethyl]-5-[(3,5-dimethoxyphenyl)methoxy]phenoxymethyl}-3,5-dimethoxybenzene
Molecular Weight (g/mol): 1047.99
Molecular Formula: C57H59BrO14
InChi Key: BMISSUNAKMQTFS-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(COC2=CC(OCC3=CC(OC)=CC(OC)=C3)=CC(COC3=CC(OCC4=CC(OCC5=CC(OC)=CC(OC)=C5)=CC(OCC5=CC(OC)=CC(OC)=C5)=C4)=CC(CBr)=C3)=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results