missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[4-(Trifluoromethyl)phenyl]propionic Acid 98.0+%, TCI America™
Supplier: TCI America T29271G
Specifications
| 3-[4-(Trifluoromethyl)phenyl]propionic Acid | |
| 115°C | |
| C10H9F3O2 | |
| 1 g | |
| OEIUMLSCWINLBB-UHFFFAOYSA-N | |
| 3-[4-(trifluoromethyl)phenyl]propanoic acid | |
| 3588736 | |
| ≥98.0% (T) |
| 53473-36-2 | |
| White-Yellow | |
| MFCD00674032 | |
| 4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid | |
| C1=CC(=CC=C1CCC(=O)O)C(F)(F)F | |
| 218.175 | |
| 218.18 | |
| Crystalline Powder |
Chemical Identifiers
| 53473-36-2 | |
| 218.175 | |
| OEIUMLSCWINLBB-UHFFFAOYSA-N | |
| 3588736 | |
| C1=CC(=CC=C1CCC(=O)O)C(F)(F)F |
| C10H9F3O2 | |
| MFCD00674032 | |
| 4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid | |
| 3-[4-(trifluoromethyl)phenyl]propanoic acid |
Safety and Handling
TSCA : No