missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Trifluoromethyl)phenyl]propionic Acid 98.0+%, TCI America™

$70.84 - $238.60

Chemical Identifiers

CAS 53473-36-2
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.175
MDL Number MFCD00674032
InChI Key OEIUMLSCWINLBB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid
PubChem CID 3588736
IUPAC Name 3-[4-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T29271G
View Documents
TCI America
T29271G
1 g
Each for $70.84
Only null left
 
T29275G
View Documents
TCI America
T29275G
5 g
Each for $238.60
Only null left
 
Description

Description

Specifications

Chemical Identifiers

53473-36-2
218.175
OEIUMLSCWINLBB-UHFFFAOYSA-N
3588736
C1=CC(=CC=C1CCC(=O)O)C(F)(F)F
C10H9F3O2
MFCD00674032
4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid
3-[4-(trifluoromethyl)phenyl]propanoic acid

Specifications

53473-36-2
White-Yellow
MFCD00674032
4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid
C1=CC(=CC=C1CCC(=O)O)C(F)(F)F
218.175
218.18
Crystalline Powder
115°C
C10H9F3O2
1 g
OEIUMLSCWINLBB-UHFFFAOYSA-N
3-[4-(trifluoromethyl)phenyl]propanoic acid
3588736
≥98.0% (T)
3-[4-(Trifluoromethyl)phenyl]propionic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents