Learn More
3-(4-Thiazolyl)-L-alanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6351103
| Quantity | 1 g |
|---|
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store at (-20°C). Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 119433-80-6 | |
| 172.202 | |
| WBZIGVCQRXJYQD-YFKPBYRVSA-N | |
| 2761495 | |
| C1=C(N=CS1)CC(C(=O)O)N |
| C6H8N2O2S | |
| MFCD00079677 | |
| 3-4-thiazolyl-l-alanine, s-2-amino-3-thiazol-4-yl propanoic acid, thiazolylalanine, 3-4-thiazoyl-l-alanine, unii-vtl1y3s44b, 2s-2-amino-3-1,3-thiazol-4-yl propanoic acid, vtl1y3s44b, s-3-4-thiazolyl alanine, l-thiazolylalanine, thiazolylalanine, l | |
| (2S)-2-amino-3-(1,3-thiazol-4-yl)propanoic acid |
Specifications
| 3-(4-Thiazolyl)-L-alanine | |
| Yellow | |
| MFCD00079677 | |
| Slightly soluble in water. | |
| C1=C(N=CS1)CC(C(=O)O)N | |
| 1 g | |
| 2761495 | |
| 95% |
| 119433-80-6 | |
| C6H8N2O2S | |
| 3-4-thiazolyl-l-alanine, s-2-amino-3-thiazol-4-yl propanoic acid, thiazolylalanine, 3-4-thiazoyl-l-alanine, unii-vtl1y3s44b, 2s-2-amino-3-1,3-thiazol-4-yl propanoic acid, vtl1y3s44b, s-3-4-thiazolyl alanine, l-thiazolylalanine, thiazolylalanine, l | |
| WBZIGVCQRXJYQD-YFKPBYRVSA-N | |
| (2S)-2-amino-3-(1,3-thiazol-4-yl)propanoic acid | |
| 172.202 | |
| 172.2 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C