missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Thiazolyl)-L-alanine, 95%, Thermo Scientific Chemicals

Catalog No. p-7055033
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63511MD 250 mg
AAH6351103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63511MD Supplier Thermo Scientific Chemicals Supplier No. H63511MD

CAS: 119433-80-6 | Molecular Formula: C6H8N2O2S | Molecular Weight: 172.202 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store at (-20°C). Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 119433-80-6
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00079677
InChI Key WBZIGVCQRXJYQD-YFKPBYRVSA-N
Synonym 3-4-thiazolyl-l-alanine, s-2-amino-3-thiazol-4-yl propanoic acid, thiazolylalanine, 3-4-thiazoyl-l-alanine, unii-vtl1y3s44b, 2s-2-amino-3-1,3-thiazol-4-yl propanoic acid, vtl1y3s44b, s-3-4-thiazolyl alanine, l-thiazolylalanine, thiazolylalanine, l
PubChem CID 2761495
IUPAC Name (2S)-2-amino-3-(1,3-thiazol-4-yl)propanoic acid
SMILES C1=C(N=CS1)CC(C(=O)O)N

Specifications

Color Yellow
Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 172.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(4-Thiazolyl)-L-alanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only