accessibility menu, dialog, popup

UserName

H2N(S)SNHOO

CAS RN 119433-80-6

IUPAC Name: (2S)-2-amino-3-(1,3-thiazol-4-yl)propanoic acid
Synonyms: thiazolylalanine
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: WBZIGVCQRXJYQD-YFKPBYRVSA-N
SMILES: N[C@@H](CC1=CSC=N1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)