missing translation for 'onlineSavingsMsg'
Learn More

3-(4-tert-Butylphenyl)isobutyraldehyde 96.0+%, TCI America™

Catalog No. B114525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B114525ML 25 mL
B1145100ML 100 mL
B1145500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B114525ML Supplier TCI America Supplier No. B114525ML

Chemical Identifiers

CAS 80-54-6
Molecular Formula C14H20O
Molecular Weight (g/mol) 204.31
MDL Number MFCD00047655
InChI Key SDQFDHOLCGWZPU-UHFFFAOYNA-N
Synonym lilial, 3-4-tert-butyl phenyl-2-methylpropanal, 3-4-tert-butylphenyl isobutyraldehyde, lilyal, lily aldehyde, 3-4-tert-butylphenyl-2-methylpropanal, 3-4-tert-butylphenyl-2-methylpropionaldehyde, benzenepropanal, 4-1,1-dimethylethyl-.alpha.-methyl, butylphenyl methylpropional, 2-4-tert-butylbenzyl propionaldehyde
PubChem CID 228987
IUPAC Name 3-(4-tert-butylphenyl)-2-methylpropanal
SMILES CC(CC1=CC=C(C=C1)C(C)(C)C)C=O

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 mL
Formula Weight 204.31
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(4-tert-Butylphenyl)isobutyraldehyde
EINECSNumber (3)-2667
RTECSNumber MW4895000
TSCA Yes