missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Pyridinedicarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America P068225G
Specifications
| 3,4-Pyridinedicarboxylic Acid | |
| 260°C | |
| MFCD00006392 | |
| 3,4-pyridinedicarboxylic acid, cinchomeronic acid, chinchomeronic acid, 3,4-pyridinedicarboxylicacid, pyridine-3,4-dicarboxylicacid, unii-7389uzw5dy, 3,4-pyridindicarbons, cinchomeronsaure, pubchem2419, 3,4-dicarboxypyridine | |
| OC(=O)C1=CC=NC=C1C(O)=O | |
| 167.12 | |
| CHEBI:46860 | |
| ≥98.0% (GC,T) |
| 490-11-9 | |
| C7H5NO4 | |
| 25 g | |
| MUYSADWCWFFZKR-UHFFFAOYSA-N | |
| pyridine-3,4-dicarboxylic acid | |
| 10273 | |
| 167.12 | |
| Crystalline Powder |
Chemical Identifiers
| 490-11-9 | |
| 167.12 | |
| MUYSADWCWFFZKR-UHFFFAOYSA-N | |
| 10273 | |
| pyridine-3,4-dicarboxylic acid |
| C7H5NO4 | |
| MFCD00006392 | |
| 3,4-pyridinedicarboxylic acid, cinchomeronic acid, chinchomeronic acid, 3,4-pyridinedicarboxylicacid, pyridine-3,4-dicarboxylicacid, unii-7389uzw5dy, 3,4-pyridindicarbons, cinchomeronsaure, pubchem2419, 3,4-dicarboxypyridine | |
| CHEBI:46860 | |
| OC(=O)C1=CC=NC=C1C(O)=O |
Safety and Handling
EINECSNumber : (5)-7026
TSCA : No