accessibility menu, dialog, popup

UserName

HOONOHO

CAS RN 490-11-9

IUPAC Name: pyridine-3,4-dicarboxylic acid
Synonyms: 3,4-pyridinedicarboxylic acid chinchomeronic acid
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: MUYSADWCWFFZKR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=NC=C1C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)