missing translation for 'onlineSavingsMsg'
Learn More

3,4-Pyridinedicarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7134726
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06825G 5 g
P068225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06825G Supplier TCI America Supplier No. P06825G

Chemical Identifiers

CAS 490-11-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006392
InChI Key MUYSADWCWFFZKR-UHFFFAOYSA-N
Synonym 3,4-pyridinedicarboxylic acid, cinchomeronic acid, chinchomeronic acid, 3,4-pyridinedicarboxylicacid, pyridine-3,4-dicarboxylicacid, unii-7389uzw5dy, 3,4-pyridindicarbons, cinchomeronsaure, pubchem2419, 3,4-dicarboxypyridine
PubChem CID 10273
ChEBI CHEBI:46860
IUPAC Name pyridine-3,4-dicarboxylic acid
SMILES OC(=O)C1=CC=NC=C1C(O)=O

Specifications

Melting Point 260°C
Quantity 5 g
Formula Weight 167.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Pyridinedicarboxylic Acid
EINECSNumber (5)-7026
TSCA No