missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Methylpiperazin-1-yl)aniline, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 148546-99-0 |
|---|---|
| Molecular Formula | C11H17N3 |
| Molecular Weight (g/mol) | 191.278 |
| InChI Key | RJGHJWKQCJAJEP-UHFFFAOYSA-N |
| Synonym | 3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline |
| PubChem CID | 11564613 |
| IUPAC Name | 3-(4-methylpiperazin-1-yl)aniline |
| SMILES | CN1CCN(CC1)C2=CC(=CC=C2)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC35914CB
|
N/A | 250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC35914DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC35914DE
|
N/A | 5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 148546-99-0 | |
| 191.278 | |
| 3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline | |
| 3-(4-methylpiperazin-1-yl)aniline |
| C11H17N3 | |
| RJGHJWKQCJAJEP-UHFFFAOYSA-N | |
| 11564613 | |
| CN1CCN(CC1)C2=CC(=CC=C2)N |
Specifications
| 148546-99-0 | |
| C11H17N3 | |
| RJGHJWKQCJAJEP-UHFFFAOYSA-N | |
| 3-(4-methylpiperazin-1-yl)aniline | |
| 11564613 | |
| 97% |
| 97% | |
| 3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline | |
| CN1CCN(CC1)C2=CC(=CC=C2)N | |
| 191.278 | |
| 191.28 | |
| 3-(4-Methylpiperazin-1-yl)aniline |