missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Methylpiperazin-1-yl)aniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC35914CB
| Quantity | 250mg |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 148546-99-0 | |
| 191.278 | |
| 3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline | |
| 3-(4-methylpiperazin-1-yl)aniline |
| C11H17N3 | |
| RJGHJWKQCJAJEP-UHFFFAOYSA-N | |
| 11564613 | |
| CN1CCN(CC1)C2=CC(=CC=C2)N |
Specifications
| 3-(4-Methylpiperazin-1-yl)aniline | |
| 97% | |
| C11H17N3 | |
| 3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline | |
| CN1CCN(CC1)C2=CC(=CC=C2)N | |
| 191.278 | |
| 191.28 |
| 148546-99-0 | |
| Amber Glass Bottle | |
| 250mg | |
| RJGHJWKQCJAJEP-UHFFFAOYSA-N | |
| 3-(4-methylpiperazin-1-yl)aniline | |
| 11564613 | |
| 97% |