missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methylpiperazin-1-yl)aniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC35914DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC35914DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

148546-99-0
191.278
3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline
3-(4-methylpiperazin-1-yl)aniline
C11H17N3
RJGHJWKQCJAJEP-UHFFFAOYSA-N
11564613
CN1CCN(CC1)C2=CC(=CC=C2)N

Specifications

3-(4-Methylpiperazin-1-yl)aniline
97%
C11H17N3
3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline
CN1CCN(CC1)C2=CC(=CC=C2)N
191.278
191.28
148546-99-0
Amber Glass Bottle
1g
RJGHJWKQCJAJEP-UHFFFAOYSA-N
3-(4-methylpiperazin-1-yl)aniline
11564613
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions