missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)propionitrile, 98%

Catalog No. AAL0793403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0793403 1 g
AAL0793406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0793403 Supplier Thermo Scientific Chemicals Supplier No. L0793403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17362-17-3 | C9H9NO | 147.18 g/mol

3-(4-Hydroxyphenyl)propionitrile is commonly used as a tool to evaluate the biological role of ERβ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Hydroxyphenyl)propionitrile is commonly used as a tool to evaluate the biological role of ERβ.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 17362-17-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00016492
InChI Key KDMJGLYRWRHKJS-UHFFFAOYSA-N
Synonym 3-4-hydroxyphenyl propionitrile, 3-4-hydroxyphenyl propanenitrile, benzenepropanenitrile, 4-hydroxy, 3-p-hydroxyphenyl propiononitrile, 3-4-hydroxy-phenyl-propionitrile, .beta.-4-hydroxyphenyl propionitrile, pubchem13636, acmc-1bvt5, 4-hydroxyhydrocinnamonitrile, 4-hydroxyphenylpropionitrile
PubChem CID 87079
IUPAC Name 3-(4-hydroxyphenyl)propanenitrile
SMILES OC1=CC=C(CCC#N)C=C1

Specifications

Melting Point 54°C to 58°C
Boiling Point 166°C (8 mmHg)
Quantity 1 g
UN Number UN3439
Beilstein 1448680
Solubility Information Insoluble in water.
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 3-(4-Hydroxyphenyl)propionitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 241-393-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only