CAS RN 17362-17-3
IUPAC Name:
3-(4-hydroxyphenyl)propanenitrile
Synonyms:
3-(4-hydroxyphenyl)propanenitrile
Molecular Weight (g/mol):
147.18
Molecular Formula:
C9H9NO
InChi Key:
KDMJGLYRWRHKJS-UHFFFAOYSA-N
SMILES:
OC1=CC=C(CCC#N)C=C1
1
–
1
of
1
results
1
Promotions Available
3-(4-Hydroxyphenyl)propionitrile, 98%
3-(4-Hydroxyphenyl)propionitrile, 98%
CAS: 17362-17-3 Molecular Formula: C9H9NO Molecular Weight (g/mol): 147.18 MDL Number: MFCD00016492 InChI Key: KDMJGLYRWRHKJS-UHFFFAOYSA-N Synonym: 3-4-hydroxyphenyl propionitrile,3-4-hydroxyphenyl propanenitrile,benzenepropanenitrile, 4-hydroxy,3-p-hydroxyphenyl propiononitrile,3-4-hydroxy-phenyl-propionitrile,.beta.-4-hydroxyphenyl propionitrile,pubchem13636,acmc-1bvt5,4-hydroxyhydrocinnamonitrile,4-hydroxyphenylpropionitrile PubChem CID: 87079 IUPAC Name: 3-(4-hydroxyphenyl)propanenitrile SMILES: OC1=CC=C(CCC#N)C=C1