missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH3209006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3209006 5 g
AAH3209003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3209006 Supplier Thermo Scientific Chemicals Supplier No. H3209006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1736-21-6 ∣ C11H8FNO3 ∣ 221.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1736-21-6
Molecular Formula C11H8FNO3
Molecular Weight (g/mol) 221.19
MDL Number MFCD03407356
InChI Key PDEGBONVUJDOFN-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-4-fluorophenyl-5-methyl, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methylisoxazol-4-carboxylic acid, buttpark 9912-50, 4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methyl, 4-isoxazolecarboxylicacid,3-4-fluorophenyl-5-methyl, 3-4-fluoro-phenyl-5-methyl-isoxazole-4-carboxylic acid
PubChem CID 2780709
IUPAC Name 3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=CC=C(F)C=C1

Specifications

Melting Point 199°C to 201°C
Quantity 5 g
Sensitivity Light and moisture sensitive
Formula Weight 221.19
Percent Purity 97%
Chemical Name or Material 3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only