accessibility menu, dialog, popup

UserName

CH3NOFOOH

CAS RN 1736-21-6

IUPAC Name: 3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Synonyms: 3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 221.19
Molecular Formula: C11H8FNO3
InChi Key: PDEGBONVUJDOFN-UHFFFAOYSA-N
SMILES: CC1=C(C(O)=O)C(=NO1)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)