missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)-5-methylisoxazol-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7130032
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10231G 1 g
F10235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10231G Supplier TCI America Supplier No. F10231G
Only null left

Chemical Identifiers

CAS 1736-21-6
Molecular Formula C11H8FNO3
Molecular Weight (g/mol) 221.19
MDL Number MFCD03407356
InChI Key PDEGBONVUJDOFN-UHFFFAOYSA-N
PubChem CID 2780709
IUPAC Name 3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=CC=C(F)C=C1

Specifications

Melting Point 201°C
Color White-Yellow
Quantity 1 g
Synonym 3-4-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-4-fluorophenyl-5-methyl, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methylisoxazol-4-carboxylic acid, buttpark 9912-50, 4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methyl, 4-isoxazolecarboxylicacid,3-4-fluorophenyl-5-methyl, 3-4-fluoro-phenyl-5-methyl-isoxazole-4-carboxylic acid
Formula Weight 221.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Fluorophenyl)-5-methylisoxazol-4-carboxylic Acid
TSCA No
Recommended Storage Refrigerator