missing translation for 'onlineSavingsMsg'
Learn More

3,4-Epoxytetrahydrofuran 97.0+%, TCI America™

Supplier:  TCI America E079525G

 View more versions of this product

Catalog No. E079525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3,4-Epoxytetrahydrofuran
Yellow
25 g
C4H6O2
1993
AIUTZIYTEUMXGG-UHFFFAOYNA-N
3,6-dioxabicyclo[3.1.0]hexane
67511
≥97.0% (GC)
285-69-8
144°C
Clear Liquid at 20°C
MFCD00800639
3,4-epoxytetrahydrofuran, 3,6-dioxabicyclo 3.1.0 hexane, 3,6-dioxa-bicyclo 3.1.0 hexane, acmc-1ccip, 3,4-epoxy-tetrahydrofuran, 3,4-diepoxytetrahydrofuran, 3,4-epoxy tetrahydrofuran
C1OCC2OC12
86.09
86.09

Chemical Identifiers

285-69-8
86.09
AIUTZIYTEUMXGG-UHFFFAOYNA-N
67511
C1OCC2OC12
C4H6O2
MFCD00800639
3,4-epoxytetrahydrofuran, 3,6-dioxabicyclo 3.1.0 hexane, 3,6-dioxa-bicyclo 3.1.0 hexane, acmc-1ccip, 3,4-epoxy-tetrahydrofuran, 3,4-diepoxytetrahydrofuran, 3,4-epoxy tetrahydrofuran
3,6-dioxabicyclo[3.1.0]hexane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions