missing translation for 'onlineSavingsMsg'
Learn More

3,4-Epoxytetrahydrofuran 97.0+%, TCI America™

Catalog No. p-7129154
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
E07955G 5 g
E079525G 25 g
2 options

Catalog No. E07955G

Supplier: TCI America E07955G

Only null left

Chemical Identifiers

CAS 285-69-8
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00800639
InChI Key AIUTZIYTEUMXGG-UHFFFAOYNA-N
Synonym 3,4-epoxytetrahydrofuran, 3,6-dioxabicyclo 3.1.0 hexane, 3,6-dioxa-bicyclo 3.1.0 hexane, acmc-1ccip, 3,4-epoxy-tetrahydrofuran, 3,4-diepoxytetrahydrofuran, 3,4-epoxy tetrahydrofuran
PubChem CID 67511
IUPAC Name 3,6-dioxabicyclo[3.1.0]hexane
SMILES C1OCC2OC12

Specifications

Color Yellow
Boiling Point 144°C
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 1993
Formula Weight 86.09
Percent Purity ≥97.0% (GC)
Chemical Name or Material 3,4-Epoxytetrahydrofuran