missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenol 98.0+%, TCI America™

Catalog No. p-7126426
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D077725G 25 g
D0777500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D077725G Supplier TCI America Supplier No. D077725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-65-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002304
InChI Key YCOXTKKNXUZSKD-UHFFFAOYSA-N
Synonym 3,4-xylenol, phenol, 3,4-dimethyl, 4,5-dimethylphenol, 3,4-dimethyl phenol, 1,3,4-xylenol, 4-hydroxy-1,2-dimethylbenzene, 1-hydroxy-3,4-dimethylbenzene, 3,4-dimethyl-phenol, 3,4-dmp, ccris 723
PubChem CID 7249
ChEBI CHEBI:39839
IUPAC Name 3,4-dimethylphenol
SMILES CC1=CC=C(O)C=C1C

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 225°C
Quantity 25 g
UN Number 2261
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethylphenol
EINECSNumber (3)-0521&(4)-0057
RTECSNumber ZE6300000
TSCA Yes