missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylhippuric Acid 98.0+%, TCI America™

Catalog No. D22415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22415G 5 g
D22411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22415G Supplier TCI America Supplier No. D22415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23082-12-4
Molecular Formula C11H12NO3
Molecular Weight (g/mol) 206.22
MDL Number MFCD00191414
InChI Key ZDHXVMSVUHHHAE-UHFFFAOYSA-M
Synonym N-(3,4-Dimethylbenzoyl)glycine
PubChem CID 152464
IUPAC Name 2-[(3,4-dimethylphenyl)formamido]acetate
SMILES CC1=CC=C(C=C1C)C(=O)NCC([O-])=O

Specifications

Melting Point 160°C
Color White
Quantity 5 g
Formula Weight 207.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethylhippuric Acid