accessibility menu, dialog, popup

UserName

CAS RN 23082-12-4

IUPAC Name: 2-[(3,4-dimethylphenyl)formamido]acetate
Synonyms: 2-[(3,4-dimethylphenyl)formamido]acetate
Molecular Weight (g/mol): 206.22
Molecular Formula: C11H12NO3
InChi Key: ZDHXVMSVUHHHAE-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1C)C(=O)NCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results