missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzeneboronic acid, 98+%

Catalog No. AAL1746103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1746103 1 g
AAL1746106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1746103 Supplier Thermo Scientific Chemicals Supplier No. L1746103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 55499-43-9 | C8H11BO2 | 149.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55499-43-9
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD01009694
InChI Key KDVZJKOYSOFXRV-UHFFFAOYSA-N
Synonym 3,4-dimethylphenyl boronic acid, 3,4-dimethylbenzeneboronic acid, 3,4-dimethylphenyl boranediol, 3,4-dimethylphenylboronicacid, 4-borono-o-xylene, boronic acid, 3,4-dimethylphenyl, pubchem1832, acmc-209lna, 3.4-dimethylbenzeneboronic acid, 3,4-dimethyl phenylboronic acid
PubChem CID 2734348
IUPAC Name (3,4-dimethylphenyl)boronic acid
SMILES CC1=CC=C(C=C1C)B(O)O

Specifications

Melting Point ∼220°C
Quantity 1 g
Beilstein 2963858
Formula Weight 149.99
Percent Purity ≥98%
Chemical Name or Material 3,4-Dimethylbenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only