missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzamide, 98%

Catalog No. p-7081823
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1199503 1 g
AAA1199506 5 g
AAA1199514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1199503 Supplier Thermo Scientific Chemicals Supplier No. A1199503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5580-33-6 | C9H11NO | 149.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5580-33-6
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00052960
InChI Key INGCXEIJXKQPJH-UHFFFAOYSA-N
PubChem CID 21755
IUPAC Name 3,4-dimethylbenzamide
SMILES CC1=C(C=C(C=C1)C(=O)N)C

Specifications

Melting Point 101°C to 103°C
Quantity 1 g
Synonym unii-1rmm93929w, benzamide, 3,4-dimethyl, acmc-20anek, 3.4-dimethylbenzamid, benzamide,3,4-dimethyl, benzamide, 3,4-dimethyl-6ci,7ci,8ci,9ci
Formula Weight 149.19
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only