accessibility menu, dialog, popup

UserName

CH3H3CNH2O

CAS RN 5580-33-6

IUPAC Name: 3,4-dimethylbenzamide
Synonyms: 3,4-dimethylbenzamide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: INGCXEIJXKQPJH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)