missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethylbenzaldehyde, 97%
Description
3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.
Solubility
Soluble in ethanol, ether, toluene. Insoluble in water.
Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.
Solubility
Soluble in ethanol, ether, toluene. Insoluble in water.
Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
Specifications
Specifications
| Density | 1.017 |
| Boiling Point | 226°C |
| Odor | Almond-like |
| Refractive Index | 1.551 |
| Quantity | 5 g |
| Beilstein | 1634270 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in ethanol,ether,toluene. Insoluble in water. |
| Formula Weight | 134.18 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| EINECSNumber | 227-770-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only