missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylacetone 97.0+%, TCI America™

Catalog No. p-7137903
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
V005225ML 25 mL
V0052500ML 500 mL
2 options

Catalog No. V005225ML

Supplier: TCI America V005225ML

Only null left

Chemical Identifiers

CAS 776-99-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008772
InChI Key UMYZWICEDUEWIM-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylacetone, 1-3,4-dimethoxyphenyl propan-2-one, 3,4-dimethoxyphenyl acetone, veratryl-2-propanone, 1-3,4-dimethoxyphenyl-2-propanone, veratryl acetone, 3,4-dimethoxybenzyl methyl ketone, 1-3,4-dimethoxyphenyl acetone, 2-propanone, 1-3,4-dimethoxyphenyl, veratryl methyl ketone
PubChem CID 69896
IUPAC Name 1-(3,4-dimethoxyphenyl)propan-2-one
SMILES COC1=CC=C(CC(C)=O)C=C1OC

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 mL
Formula Weight 194.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethoxyphenylacetone
EINECSNumber (8)-0683
RTECSNumber UC1795500
TSCA No