Learn More
3,4-Dimethoxyphenol, 98%
CAS: 2033-89-8 | C8H10O3 | 154.165 g/mol
$2,508.49 - $2,508.49
Chemical Identifiers
| CAS | 2033-89-8 |
|---|---|
| Molecular Formula | C8H10O3 |
| Molecular Weight (g/mol) | 154.165 |
| MDL Number | MFCD00008390 |
| InChI Key | SMFFZOQLHYIRDA-UHFFFAOYSA-N |
| Synonym | phenol, 3,4-dimethoxy, 4-hydroxyveratrole, 3,4-bis methyloxy phenol, unii-38b43wcu83, 3,4-dimethoxy phenol, 3,4-dimethoxyphel, 3,4,dimethoxyphenol, phenol,4-dimethoxy, 3,4-dimethoxy-pheno, pubchem4113 |
| PubChem CID | 16251 |
| IUPAC Name | 3,4-dimethoxyphenol |
| SMILES | COC1=C(C=C(C=C1)O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1134422
|
Thermo Scientific Chemicals
A1134422 |
100 g |
N/A
|
|
|||||
Description
3,4-Dimethoxyphenol is used in the preparation of 5,6-dimethoxy benzofuranone derivatives and multi-target anti Alzheimer compounds. It is also used in the preparation of 3,4-dimethoxyphenyl-beta-D-glucopyranoside and 4-(but-2-enyloxy)-1,2-dimethoxybenzene. Further, it serves as a precursor for 4H-chromenes synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dimethoxyphenol is used in the preparation of 5,6-dimethoxy benzofuranone derivatives and multi-target anti Alzheimer compounds. It is also used in the preparation of 3,4-dimethoxyphenyl-beta-D-glucopyranoside and 4-(but-2-enyloxy)-1,2-dimethoxybenzene. Further, it serves as a precursor for 4H-chromenes synthesis.
Solubility
Soluble in water.
Notes
Light sensitive. Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.
Chemical Identifiers
| 2033-89-8 | |
| 154.165 | |
| SMFFZOQLHYIRDA-UHFFFAOYSA-N | |
| 16251 | |
| COC1=C(C=C(C=C1)O)OC |
| C8H10O3 | |
| MFCD00008390 | |
| phenol, 3,4-dimethoxy, 4-hydroxyveratrole, 3,4-bis methyloxy phenol, unii-38b43wcu83, 3,4-dimethoxy phenol, 3,4-dimethoxyphel, 3,4,dimethoxyphenol, phenol,4-dimethoxy, 3,4-dimethoxy-pheno, pubchem4113 | |
| 3,4-dimethoxyphenol |
Specifications
| 2033-89-8 | |
| 167°C (14 mmHg) | |
| MFCD00008390 | |
| 1366650 | |
| phenol, 3,4-dimethoxy, 4-hydroxyveratrole, 3,4-bis methyloxy phenol, unii-38b43wcu83, 3,4-dimethoxy phenol, 3,4-dimethoxyphel, 3,4,dimethoxyphenol, phenol,4-dimethoxy, 3,4-dimethoxy-pheno, pubchem4113 | |
| SMFFZOQLHYIRDA-UHFFFAOYSA-N | |
| 3,4-dimethoxyphenol | |
| 16251 | |
| 98% |
| 79°C to 82°C | |
| C8H10O3 | |
| 100 g | |
| Light sensitive | |
| Soluble in water. | |
| COC1=C(C=C(C=C1)O)OC | |
| 154.165 | |
| 154.16 | |
| 3,4-Dimethoxyphenol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-995-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only