missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxyphenol, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 77.0°C to 82.0°C |
| Color | White to Beige |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | (CH3O)2C6H3OH |
| Quantity | 10 g |
| Beilstein | 06,III,6277 |
| Formula Weight | 154.17 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| EINECSNumber | 217-995-4 |
| Recommended Storage | Product may darken during storage |
RUO – Research Use Only