missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzylamine 97.0+%, TCI America™

Catalog No. p-7128058
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40245G 5 g
D402425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40245G Supplier TCI America Supplier No. D40245G
Only null left

Chemical Identifiers

CAS 5763-61-1
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00008116
InChI Key DIVNUTGTTIRPQA-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzylamine, veratrylamine, 3,4-dimethoxyphenyl methanamine, benzenemethanamine, 3,4-dimethoxy, 1-3,4-dimethoxyphenyl methanamine, benzylamine, 3,4-dimethoxy, 3,4-dimethoxyphenyl methylamine, veratrylamin, pubchem7394, acmc-1altc
PubChem CID 79832
IUPAC Name 1-(3,4-dimethoxyphenyl)methanamine
SMILES COC1=CC=C(CN)C=C1OC

Specifications

Color Yellow
Boiling Point 156°C
Quantity 5 g
UN Number 2735
Formula Weight 167.21
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethoxybenzylamine
RTECSNumber YX7355000
TSCA No