missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimercaptotoluene, 95%

Catalog No. p-113361
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421180010 1 g Glass bottle
AC421180050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC421180010 Supplier Thermo Scientific Chemicals Supplier No. 421180010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 496-74-2 | C7H8S2 | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 496-74-2
Molecular Formula C7H8S2
Molecular Weight (g/mol) 156.27
MDL Number MFCD00004844
InChI Key NIAAGQAEVGMHPM-UHFFFAOYSA-N
Synonym toluene-3,4-dithiol, dithiol, 1,2-benzenedithiol, 4-methyl, 3,4-dimercaptotoluene, 3,4-dimercaptotoluol, 1,2-dimercapto-4-methylbenzene, 4-methyl-1,2-benzenedithiol, 3,4-toluenedithiol, o-toluenesulfonylamide, usaf b-59
PubChem CID 10334
IUPAC Name 4-methylbenzene-1,2-dithiol
SMILES CC1=CC(=C(C=C1)S)S

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 27°C
Color White to Yellow
Density 1.1800g/mL
Boiling Point 185°C (84.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.18
Merck Index 15, 9690
Formula Weight 156.27
Percent Purity 95%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 3, 4-Dimercaptotoluene, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-828-3
RTECSNumber XT2275000
TSCA TSCA

RUO – Research Use Only