Learn More
3,4-Dihydroxyphenylacetic acid, 98+%
CAS: 102-32-9 | C8H8O4 | 168.148 g/mol
Supplier: Thermo Scientific Chemicals A1589306
Description
3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.
Solubility
Soluble in water and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Air sensitive. Incompatible with strong bases and strong oxidizing agents. Store at room temperature
Specifications
| 3,4-Dihydroxyphenylacetic acid | |
| 128°C to 130°C | |
| MFCD00004338 | |
| 2211017 | |
| Soluble in water and methanol. | |
| C1=CC(=C(C=C1CC(=O)O)O)O | |
| 168.148 | |
| CHEBI:41941 | |
| ≥98% |
| 102-32-9 | |
| C8H8O4 | |
| 5 g | |
| 3,4-dihydroxyphenylacetic acid, dopac, homoprotocatechuic acid, 2-3,4-dihydroxyphenyl acetic acid, dopacetic acid, benzeneacetic acid, 3,4-dihydroxy, dihydroxyphenylacetic acid, 3,4-dihydroxybenzeneacetic acid, 4-carboxymethylcatechol, homoprotocatechuate | |
| CFFZDZCDUFSOFZ-UHFFFAOYSA-N | |
| 2-(3,4-dihydroxyphenyl)acetic acid | |
| 547 | |
| 168.15 |
Chemical Identifiers
| 102-32-9 | |
| 168.148 | |
| CFFZDZCDUFSOFZ-UHFFFAOYSA-N | |
| 547 | |
| 2-(3,4-dihydroxyphenyl)acetic acid |
| C8H8O4 | |
| MFCD00004338 | |
| 3,4-dihydroxyphenylacetic acid, dopac, homoprotocatechuic acid, 2-3,4-dihydroxyphenyl acetic acid, dopacetic acid, benzeneacetic acid, 3,4-dihydroxy, dihydroxyphenylacetic acid, 3,4-dihydroxybenzeneacetic acid, 4-carboxymethylcatechol, homoprotocatechuate | |
| CHEBI:41941 | |
| C1=CC(=C(C=C1CC(=O)O)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 203-024-1
RTECSNumber : AH0590000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only