missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxyphenylacetic acid, 98+%

Catalog No. AAA1589303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1589303 1 g
AAA1589306 5 g
AAA1589314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1589303 Supplier Thermo Scientific Chemicals Supplier No. A1589303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 102-32-9 | C8H8O4 | 168.148 g/mol

3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.

Solubility
Soluble in water and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Air sensitive. Incompatible with strong bases and strong oxidizing agents. Store at room temperature

Chemical Identifiers

CAS 102-32-9
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00004338
InChI Key CFFZDZCDUFSOFZ-UHFFFAOYSA-N
Synonym 3,4-dihydroxyphenylacetic acid, dopac, homoprotocatechuic acid, 2-3,4-dihydroxyphenyl acetic acid, dopacetic acid, benzeneacetic acid, 3,4-dihydroxy, dihydroxyphenylacetic acid, 3,4-dihydroxybenzeneacetic acid, 4-carboxymethylcatechol, homoprotocatechuate
PubChem CID 547
ChEBI CHEBI:41941
IUPAC Name 2-(3,4-dihydroxyphenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)O)O

Specifications

Melting Point 128°C to 130°C
Quantity 1 g
Beilstein 2211017
Solubility Information Soluble in water and methanol.
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material 3,4-Dihydroxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-024-1
RTECSNumber AH0590000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only