missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7126660
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D128325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D128325G Supplier TCI America Supplier No. D128325G
Only null left

Chemical Identifiers

CAS 102-32-9
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00004338
InChI Key CFFZDZCDUFSOFZ-UHFFFAOYSA-N
Synonym 3,4-dihydroxyphenylacetic acid, dopac, homoprotocatechuic acid, 2-3,4-dihydroxyphenyl acetic acid, dopacetic acid, benzeneacetic acid, 3,4-dihydroxy, dihydroxyphenylacetic acid, 3,4-dihydroxybenzeneacetic acid, 4-carboxymethylcatechol, homoprotocatechuate
PubChem CID 547
ChEBI CHEBI:41941
IUPAC Name 2-(3,4-dihydroxyphenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)O)O

Specifications

Melting Point 130°C
Quantity 25 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydroxyphenylacetic Acid
RTECSNumber AH0590000
TSCA No