missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydroxybenzonitrile 98.0+%, TCI America™
$57.79 - $233.00
Chemical Identifiers
| CAS | 17345-61-8 |
|---|---|
| Molecular Formula | C7H5NO2 |
| Molecular Weight (g/mol) | 135.122 |
| MDL Number | MFCD00016436 |
| InChI Key | NUWHYWYSMAPBHK-UHFFFAOYSA-N |
| Synonym | protocatachuonitrile, 3,4-dihydroxy benzonitrile, 3,4-dihydroxybenzenecarbonitrile, protocatechuonitrile, 4-cyanocatechol, protocatechunitrile, 4-cyanopyrocatechol, pubchem3120, acmc-209e6q, 3, 4-dihydroxybenzonitrile |
| PubChem CID | 87065 |
| IUPAC Name | 3,4-dihydroxybenzonitrile |
| SMILES | C1=CC(=C(C=C1C#N)O)O |
Chemical Identifiers
| 17345-61-8 | |
| 135.122 | |
| NUWHYWYSMAPBHK-UHFFFAOYSA-N | |
| 87065 | |
| C1=CC(=C(C=C1C#N)O)O |
| C7H5NO2 | |
| MFCD00016436 | |
| protocatachuonitrile, 3,4-dihydroxy benzonitrile, 3,4-dihydroxybenzenecarbonitrile, protocatechuonitrile, 4-cyanocatechol, protocatechunitrile, 4-cyanopyrocatechol, pubchem3120, acmc-209e6q, 3, 4-dihydroxybenzonitrile | |
| 3,4-dihydroxybenzonitrile |
Specifications
| 17345-61-8 | |
| White-Yellow | |
| MFCD00016436 | |
| 3439 | |
| NUWHYWYSMAPBHK-UHFFFAOYSA-N | |
| 3,4-dihydroxybenzonitrile | |
| 87065 | |
| ≥98.0% (GC) | |
| 3,4-Dihydroxybenzonitrile |
| 157°C | |
| C7H5NO2 | |
| 5 g | |
| protocatachuonitrile, 3,4-dihydroxy benzonitrile, 3,4-dihydroxybenzenecarbonitrile, protocatechuonitrile, 4-cyanocatechol, protocatechunitrile, 4-cyanopyrocatechol, pubchem3120, acmc-209e6q, 3, 4-dihydroxybenzonitrile | |
| C1=CC(=C(C=C1C#N)O)O | |
| 135.122 | |
| 135.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No