accessibility menu, dialog, popup

UserName

OHHON

CAS RN 17345-61-8

IUPAC Name: 3,4-dihydroxybenzonitrile
Synonyms: 3,4-dihydroxybenzonitrile
Molecular Weight (g/mol): 135.12
Molecular Formula: C7H5NO2
InChi Key: NUWHYWYSMAPBHK-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1O)C#N
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)