missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydroxybenzoic Acid 98.0+%, TCI America™
Supplier: TCI America C0055100G
Specifications
| 3,4-Dihydroxybenzoic Acid | |
| 199°C | |
| MFCD00002509 | |
| protocatechuic acid, 4-carboxy-1,2-dihydroxybenzene, protocatehuic acid, benzoic acid, 3,4-dihydroxy, protocatechuate, 4,5-dihydroxybenzoic acid, 3,4-dihydroxybenzoicacid, unii-36r5qj8l4b, ccris 6291, chembl37537 | |
| C1=CC(=C(C=C1C(=O)O)O)O | |
| 154.121 | |
| CHEBI:36062 | |
| ≥98.0% (HPLC) |
| 99-50-3 | |
| C7H6O4 | |
| 100 g | |
| YQUVCSBJEUQKSH-UHFFFAOYSA-N | |
| 3,4-dihydroxybenzoic acid | |
| 72 | |
| 154.12 | |
| Crystalline Powder |
Chemical Identifiers
| 99-50-3 | |
| 154.121 | |
| YQUVCSBJEUQKSH-UHFFFAOYSA-N | |
| 72 | |
| 3,4-dihydroxybenzoic acid |
| C7H6O4 | |
| MFCD00002509 | |
| protocatechuic acid, 4-carboxy-1,2-dihydroxybenzene, protocatehuic acid, benzoic acid, 3,4-dihydroxy, protocatechuate, 4,5-dihydroxybenzoic acid, 3,4-dihydroxybenzoicacid, unii-36r5qj8l4b, ccris 6291, chembl37537 | |
| CHEBI:36062 | |
| C1=CC(=C(C=C1C(=O)O)O)O |
Safety and Handling
EINECSNumber : (3)-1616
RTECSNumber : UL0560000
TSCA : No