missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. C005525G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C005525G 25 g
C0055100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C005525G Supplier TCI America Supplier No. C005525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-50-3
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002509
InChI Key YQUVCSBJEUQKSH-UHFFFAOYSA-N
Synonym protocatechuic acid, 4-carboxy-1,2-dihydroxybenzene, protocatehuic acid, benzoic acid, 3,4-dihydroxy, protocatechuate, 4,5-dihydroxybenzoic acid, 3,4-dihydroxybenzoicacid, unii-36r5qj8l4b, ccris 6291, chembl37537
PubChem CID 72
ChEBI CHEBI:36062
IUPAC Name 3,4-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)O)O

Specifications

Melting Point 199°C
Quantity 25 g
Formula Weight 154.12
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydroxybenzoic Acid
EINECSNumber (3)-1616
RTECSNumber UL0560000
TSCA No