missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzoic acid, 97%

Catalog No. p-112315
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114890250 25 g Glass bottle
AC114891000 100 g Glass bottle
AC114895000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC114890250 Supplier Thermo Scientific Chemicals Supplier No. 114890250
Only null left

CAS: 99-50-3 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-50-3
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002509
InChI Key YQUVCSBJEUQKSH-UHFFFAOYSA-N
Synonym protocatechuic acid, 4-carboxy-1,2-dihydroxybenzene, protocatehuic acid, benzoic acid, 3,4-dihydroxy, protocatechuate, 4,5-dihydroxybenzoic acid, 3,4-dihydroxybenzoicacid, unii-36r5qj8l4b, ccris 6291, chembl37537
PubChem CID 72
ChEBI CHEBI:36062
IUPAC Name 3,4-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 198.0°C to 202.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (HO)2C6H3CO2H
Quantity 25 g
Beilstein 10,389
Merck Index 15,8003
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol and ether
Formula Weight 154.12
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3, 4-Dihydroxybenzoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-760-0
RTECSNumber UL0560000
TSCA TSCA

RUO – Research Use Only