Learn More
Thermo Scientific Chemicals 3,4-Dihydroxy-DL-phenylalanine, 98%
CAS: 63-84-3 | C9H11NO4 | 197.19 g/mol
$164.16 - $682.45
Chemical Identifiers
| CAS | 63-84-3 |
|---|---|
| Molecular Formula | C9H11NO4 |
| Molecular Weight (g/mol) | 197.19 |
| MDL Number | MFCD00063060 |
| InChI Key | WTDRDQBEARUVNC-UHFFFAOYNA-N |
| Synonym | dl-dopa, 3,4-dihydroxy-dl-phenylalanine, 2-amino-3-3,4-dihydroxyphenyl propanoic acid, dl-dioxyphenylalanine, dl-dihydroxyphenylalanine, 3-3,4-dihydroxyphenyl-dl-alanine, 3-hydroxytyrosine, 3-hydroxy-dl-tyrosine, dopa, dl-3,4-dopa |
| PubChem CID | 836 |
| ChEBI | CHEBI:49168 |
| IUPAC Name | 2-amino-3-(3,4-dihydroxyphenyl)propanoic acid |
| SMILES | NC(CC1=CC=C(O)C(O)=C1)C(O)=O |
Description
3,4-Dihydroxy-DL-phenylalanine is used as a dopamine precursor. It is also useful in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C9H11NO4 | |
| MFCD00063060 | |
| dl-dopa, 3,4-dihydroxy-dl-phenylalanine, 2-amino-3-3,4-dihydroxyphenyl propanoic acid, dl-dioxyphenylalanine, dl-dihydroxyphenylalanine, 3-3,4-dihydroxyphenyl-dl-alanine, 3-hydroxytyrosine, 3-hydroxy-dl-tyrosine, dopa, dl-3,4-dopa | |
| CHEBI:49168 | |
| NC(CC1=CC=C(O)C(O)=C1)C(O)=O |
Specifications
| 290°C (decomposition) | |
| C9H11NO4 | |
| 14,3420 | |
| Soluble in water,0.5M dilute hydrochloric acid and formic acid. Insoluble in ethanol,benzene,chloroform and ethyl acetate. | |
| NC(CC1=CC=C(O)C(O)=C1)C(O)=O | |
| 197.19 | |
| CHEBI:49168 | |
| 197.19 | |
| 98% | |
| 3,4-Dihydroxy-DL-phenylalanine |
| 63-84-3 | |
| MFCD00063060 | |
| dl-dopa, 3,4-dihydroxy-dl-phenylalanine, 2-amino-3-3,4-dihydroxyphenyl propanoic acid, dl-dioxyphenylalanine, dl-dihydroxyphenylalanine, 3-3,4-dihydroxyphenyl-dl-alanine, 3-hydroxytyrosine, 3-hydroxy-dl-tyrosine, dopa, dl-3,4-dopa | |
| WTDRDQBEARUVNC-UHFFFAOYNA-N | |
| 2-amino-3-(3,4-dihydroxyphenyl)propanoic acid | |
| 836 | |
| Odorless | |
| Air Sensitive | |
| Powder |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 200-566-0
RTECSNumber : AY5250000
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only