accessibility menu, dialog, popup

UserName

CAS RN 63-84-3

IUPAC Name: 2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
Synonyms: 3',4'-dihydroxyphenylalanine|helfo dopa|3-hydroxytyrosine|dopicar|biodopa|cerepap|DL-dihydroxyphenylalanine|sobiodopa
Molecular Weight (g/mol): 197.19
Molecular Formula: C9H11NO4
InChi Key: WTDRDQBEARUVNC-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(O)C(O)=C1)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)