missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3,4-Dihydroxy-DL-phenylalanine, 98%

Catalog No. AA4153506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4153506 5 g
AA4153514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4153506 Supplier Thermo Scientific Chemicals Supplier No. 4153506

CAS: 63-84-3 | C9H11NO4 | 197.19 g/mol

3,4-Dihydroxy-DL-phenylalanine is used as a dopamine precursor. It is also useful in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63-84-3
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00063060
InChI Key WTDRDQBEARUVNC-UHFFFAOYNA-N
Synonym dl-dopa, 3,4-dihydroxy-dl-phenylalanine, 2-amino-3-3,4-dihydroxyphenyl propanoic acid, dl-dioxyphenylalanine, dl-dihydroxyphenylalanine, 3-3,4-dihydroxyphenyl-dl-alanine, 3-hydroxytyrosine, 3-hydroxy-dl-tyrosine, dopa, dl-3,4-dopa
PubChem CID 836
ChEBI CHEBI:49168
IUPAC Name 2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=C(O)C(O)=C1)C(O)=O

Specifications

Melting Point 290°C (decomposition)
Merck Index 14,3420
Solubility Information Soluble in water,0.5M dilute hydrochloric acid and formic acid. Insoluble in ethanol,benzene,chloroform and ethyl acetate.
Quantity 5 g
Odor Odorless
Formula Weight 197.19
Sensitivity Air Sensitive
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3,4-Dihydroxy-DL-phenylalanine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-566-0
RTECSNumber AY5250000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only